Cynomolgus monkey Recombinant IL-17F Protein (from HEK293 cells)
Catalog #: 77125-094
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:102118959
- Packaging:Vial
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Lyophilized protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Protein synonyms:IL-17F|Interleukin-17F|Cytokine ML-1
- Protein/peptide name:IL17F
- Purity:>95% as determined by SDS-PAGE
- Molecular weight:16.8 kDa
- Sequence:Arg 31 - Gln 163
- Formulation:Lyophilized. PBS, pH 7.4.
- Shipping temperature:Room temperature
- Tested applications:ELISA,WB
Specifications
About this item
Customers Who Bought This Also Bought