Cynomolgus monkey Recombinant GUCY2C/Guanylyl cyclase C Protein (from HEK293 cells)
Catalog #: 77129-194
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:102130850
- Packaging:Vial
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Lyophilized protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Protein synonyms:GUCY2C|hSTAR|STA receptor|GC-C|STAR|GUC2C
- Protein/peptide name:GUCY2C
- Purity:>90% as determined by SDS-PAGE
- Molecular weight:72.6 kDa
- Sequence:Ser 24 - Gln 430
- Formulation:Lyophilized. PBS, pH 7.4.
- Shipping temperature:Room temperature
- Tested applications:ELISA,WB
Specifications
About this item
Customers Who Bought This Also Bought