Human Recombinant Netrin receptor DCC/DCC Protein (from HEK293 cells)
Catalog #: 77125-194
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:1630
- Packaging:Vial
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Lyophilized protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Protein synonyms:Netrin receptor DCC|IGDCC1|netrin receptor DCC|DCC
- Protein/peptide name:DCC
- Purity:>90% as determined by SDS-PAGE
- Molecular weight:144.1 kDa
- Sequence:Phe 32 - Asn 1097
- Formulation:Lyophilized. 50 mM Tris, 100 mM Glycine, 25 mM Arginine, 150 mM NaCl, pH 7.5.
- Shipping temperature:Room temperature
- Tested applications:WB
Specifications
About this item
Customers Who Bought This Also Bought