SARS-CoV-2 (COVID-19) Recombinant Nucleocapsid Protein (from E. coli)
Catalog #: 77133-092
Supplier: Prosci
New Product
CAS Number:
Specifications
- Packaging:Vial
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:SARS-CoV-2
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2...8 °C for one week. Store at −20...−80 °C for twelve months from the date of receipt.
- Protein synonyms:Nucleoprotein
- Protein/peptide name:Nucleocapsid
- Purity:>90% as determined by SDS-PAGE quantitative densitometry by Coomassie Blue Staining
- Molecular weight:47.79 kDa
- Sequence:Met1-Ala419
- Formulation:Lyophilized. Supplied as Lyophilized from PBS, pH 7.5. Please reconstitute the protein with ddH₂O to the concentration with 1 mg/ml.
- Shipping temperature:Room temperature
- Tested applications:ELISA,WB
Specifications
About this item
Customers Who Bought This Also Bought