Butane Burner Hot Plate
Catalog #: 470352-978
Supplier: G.S.C. INTERNATIONAL, INC.
New Product
Specifications
- Description:Butane burner hot plate
- Cat. no.:470352-978
Specifications
About this item
Unique, compact, and affordable staple for your lab!
- The Butane Hotplate is has a low profile and wide base which provides a stable footing
- The concentric rings support various sizes of flasks
- The run time when full is approximately 25 minutes
The portable butane burner is a great option for any lab because the wide base and low profile provides the burner with a stable footing. The support rings attached at the top allow you to use the burner as a hotplate for laboratory glassware. The valve and ignition buttons are separate for added safety. The flame can reach a maximum temperature of approximately 1300 decrees Celsius. The burner can be refilled quickly and easily and holds approximately 30 grams of butane. The operating time is estimated to be 25 minutes. The product is shipped without fuel.
Caution: Prop 65 Warning Labels are applied for lead.
Customers Who Bought This Also Bought