Anti-S Recombinant Antibody [Clone: Sb#16]
Catalog #: 76630-274
Supplier: Prosci
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:surface glycoprotein
- Clonality:Recombinant
- Clone:Sb#16
- Gene ID:43740568
- Isotype:IgG-Fc Fusion
- Reactivity:Virus
- Antigen symbol:S
- Conjugation:Unconjugated
- ELISA:Yes
- Size:0.2 mg
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:SARS coronavirus 2 Spike Protein|SARS-COV-2 Spike glycoprotein|Spike glycoprotein|Sb16|E2|Receptor Binding Domain|SARSCoV2|Peplomer protein|Human coronavirus 2 spike glycoprotein|SARS-CoV-2 S protein|SARSCOV2 Spike protein|SARS coronavirus 2 S protein|Severe acute respiratory syndrome virus 2 spike glycoprotein|SARS-COV-2 S protein|SARS CoV 2 Spike protein|Severe acute respiratory syndrome 2 spike glycoprotein|S glycoprotein|SARS CoV 2|SARS CoV 2 S glycoprotein|SARS-CoV2|E2 glycoprotein|RBD|2019-nCoV|COVID-19 Spike protein
- Modification:Unmodified
- Storage buffer:PBS with 0.02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This clone was originally isolated in a form of a synthetic nanobody (sybody) via a 'target swap' selection procedure against RBD-vYFP using ribosomal display and against RBD-Fc fusion during phage display rounds.
- Tested applications:Inhibition assay, grating-coupled interferometry, therapeutics
- Cat. no.:76630-274
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: surface glycoprotein
Clonality: Recombinant
Clone: Sb#16
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG-Fc Fusion
Reactivity: Virus
Customers Who Bought This Also Bought