Anti-FGB Recombinant Antibody [Clone: 8E5]
Catalog #: 76629-898
Supplier: Prosci
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:HGNC:HGNC:3662
- Clonality:Recombinant
- Clone:8E5
- Gene ID:2244
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:FGB
- Conjugation:Unconjugated
- ELISA:Yes
- Size:0.2 mg
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:factor-1a|fibrin beta-chain|Fibrinopeptide B|Factor Ia|fibrin β chain|fibrin|fibrin beta chain
- Modification:Unmodified
- Storage buffer:PBS with 0.02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:8E5 was prepared fusing Sp2/o myeloma cells with spleen cells derived from female BALB/c mice immunized with a synthetic β-fibrin peptide seqeunce GHRPLDKC conjugated to MB-KLH (maleimidobenzoylated keyhole limpet hemocyanin).
- Tested applications:In vivo, ex vivo, imaging
- Cat. no.:76629-898
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: HGNC:HGNC:3662
Clonality: Recombinant
Clone: 8E5
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1 kappa
Reactivity: Human
Customers Who Bought This Also Bought