Anti-CD3E Recombinant Antibody [Clone: YTH 12.5]
Catalog #: 76630-640
Supplier: Prosci
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD3 epsilon subunit of T-cell receptor complex
- Clonality:Recombinant
- Clone:YTH 12.5
- Gene ID:916
- Isotype:IgG lambda
- Reactivity:Human
- Antigen symbol:CD3E
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:0.2 mg
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:CD3E|CD3e|T3E|T-cell surface glycoprotein CD3 epsilon chain|T-cell surface antigen T3/Leu-4 epsilon chain|YTH12.5
- Modification:Unmodified
- Storage buffer:PBS with 0.02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Human CD3.
- Cat. no.:76630-640
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: CD3 epsilon subunit of T-cell receptor complex
Clonality: Recombinant
Clone: YTH 12.5
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG lambda
Reactivity: Human
Customers Who Bought This Also Bought