Anti-Ms4a1 Recombinant Antibody [Clone: 18B12]
Catalog #: 76630-176
Supplier: Prosci
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:membrane-spanning 4-domains, subfamily A, member 1
- Clonality:Recombinant
- Clone:18B12
- Gene ID:12482
- Isotype:IgG1 kappa
- Reactivity:Mouse
- Antigen symbol:Ms4a1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoFluorescence:Yes
- Size:0.2 mg
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:Membrane-spanning 4-domains subfamily A member 1|subfamily A|Ly-44|B-cell differentiation antigen Ly-44|CD20 antigen|CD20|member 2|Lymphocyte antigen 44|membrane-spanning 4-domains|B-lymphocyte antigen CD20
- Modification:Unmodified
- Storage buffer:PBS with 0.02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was generated by immunizing CD20−/− mice with CD20-transfected cell lines (300.18 and 70Z3) and CD20-peptide-keyhole limpet hemocyanin conjugate.
- Tested applications:Depletion,Functional Assays
- Cat. no.:76630-176
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: membrane-spanning 4-domains, subfamily A, member 1
Clonality: Recombinant
Clone: 18B12
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG1 kappa
Reactivity: Mouse
Customers Who Bought This Also Bought