Inverted Microscope with Flexacam C1, DMi1
Catalog #: CA76538-912
Supplier: Leica Microsystems
Specifications
- Description:Inverted tissue culture microscope with camera
- Cat. no.:CA76538-912
Specifications
About this item
Microscope for basic cell culture applications and imaging.
- Large dynamic range - provides true-to-life colors
- 12 MP CMOS sensor with 1.55×1.55 pixel size - reveals fine details in acquired images
- Fast auto exposure - allows quick screening of samples with bright and dark areas
- Save images to your local network or send as email attachment for easy sharing and storage
View, record, and share in seconds - take the advantage of the DMi1, an easy-to-use microscope that requires minimal training and maintenance, equipped with the FLEXACAM C1, which allows you to view images in full HD or 4K and record videos with up to 30 frames per second.
Delivery: This package contains Leica DMi1 with LED illumination; FOV 20 with an interface for exchangeable eyepieces; two eyepieces (10×/20 eyepiece with eye guard and 10×/20 focusing eyepiece with eye guard); S40 condenser; 4 position slider for S40 condenser (BF, Ph0, Ph1, Ph2); 4 objectives (HI PLAN 4×/0.10 PH0, Hi plan Ph1 (10×, 20×, 40×); dust cover fix stage (stage insert 20 mm is included); FLEXACAM C1, 12 M pixels, multiple ports (HDMI, USB, Ethernet); power cable; stand alone kit for FLEXACAM C1: USB stick, wireless mouse (USB dongle), power supply, HDMI cable, country specific power cord required.
Customers Who Bought This Also Bought