RightCare Class 100 Hand-Specific 12" Sterile Cleanroom Nitrile Gloves
Supplier: OUTSOURCE PHARMACY SOLUTIONS, INC.
New Product
Certificates
About this item
RightCare Class 100 hand specific nitrile gloves are ideal for cleanroom environments or higher controlled environments. They provide strong, reliable, durable, sensitive, and dextrous hand protection with consistent cleanliness for operators.
- Protective cleanroom nitrile gloves manufactured with no latex proteins
- Hand-specific fit with beaded and folded cuffs for aseptic donning
- Qualified for use in the pharmaceutical industry
- Particle and extractable testing in accordance with IES-RP-CC-005.2
- Gamma irradiated minimum dose 2.5 Mrads
- Superior strength offers puncture-resistant protection
- Leak-tested to ensure snug fit
The gloves are sterilized by gamma radiation to ensure they are ready for use by workers and technicians in industries such as BioTech, Electronics, Medical Device, Semiconductors, Pharmaceutical, Manufacturing, Research Laboratories, and more. Recommended for Class 100/ISO 5 Cleanrooms.
Certifications: Meets or exceeds ASTM D3578-05 Standards. Complies with US Government CFR 177.2600 for food handling. Qualified for use in the pharmaceutical industry. Certified sterile. Certificates of Radiation, Conformance, and Analysis are available.
Packaging: 200 individually packaged pairs.
Specifications
- Catalog No:
- 77148-100
- 77148-098
- 77148-096
- 77148-090
- 77148-092
- 77148-094
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Cleanroom class:
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- Material:
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Sterility:
- Sterile
- Sterile
- Sterile
- Sterile
- Sterile
- Sterile
Specifications
Related Information
Certificates
Search by Lot Number77148-090_EP-1324 CoC_Outsource_Rightcare Class 100 Hand Specific Gamma Sterilized Cleanroom 12" Nitrile Gloves
Conformity declaration
Spec Hand Specific Gamma Sterilized Cleanroom Nitrile Gloves
Product specification
Customers Who Bought This Also Bought