Specifications
- Clothing Size:One size
- Color:White
- Cat. no.:77148-082
Specifications
About this item
RightCare disposable beard covers measure 18×9" and provide full coverage over mustache, beard, and sideburns.
- High-quality breathable spunbond polypropylene
- Lightweight yet strong; ideal for everyday use while at work
- Comfortable, non-binding elastic band is latex-free, and holds the cover securely over the beard/jaw area
- Ear loop attachments make it easy to apply and remove
- Nonsterile
Facial hair can harbor bacteria, shed, and may introduce contamination into products or environments. Disposable beard covers reduce the likeliness of small hair fibers encountering clean items during food processing and packaging, food preparation, laboratories, hospitals, production areas, or within any environment sensitive to trace contamination. They are also commonly used for various environments such as restaurants, cleanroom, clinical settings, medical care facilities, law enforcement, tattoo, product manufacturing, and pharmacies.
Spunbond Polypropylene is a textile manufacturing process that improves breathability and reduces water absorption. This makes wearing the beard cover exceptionally comfortable and easier to breathe through. The beard cover also helps reduce outbound saliva, sweat, and particles from the mouth and beard.