Specifications
- Clothing Size:One size
- Color:White
- Cat. no.:77148-080
Specifications
About this item
RightCare 24" Bouffant caps provide basic protection against dirt, contaminants, and particulates in non hazardous environments.
- White, non sterile bouffant caps
- Highly breathable mesh fabric (10 to 12 g/m² spunbond polypropylene)
- Caps provide generous airflow to keep the head cool
- Dual elastic band is latex-free to reduce irritations
RightCare 24" Bouffant Caps are comfortable for workers to wear on a daily routine because they allows generous airflow to keep the head cool. Dual elastic band is latex-free and stretches easily to place the cap over the head for a snug fit. They are also effective at keeping hair contained to avoid contamination when used in various industries such as restaurants, food service, clean room environments, clinical settings, medical care facilities, tattoo product manufacturing, laboratories, hospitals, and pharmacies. Bouffant caps are also necessary to prevent cross-contamination for businesses that produce or package consumer products, pharmaceuticals, or assemble electronics.