Specifications
- Description:Model BA410E with FITC and TRITC filter fluorescence bundle
- Model:Model BA410E
- Cat. no.:CA77147-438
- Focal Length:Model BA410E with FITC and TRITC filter fluorescence bundle
Specifications
About this item
This new fluorescence bundle includes BA410E 3WLED trinocular sextuple microscope (light split 80/20), complete EPI-fluorescence attachment, EGFP/FITC/Cy2/AlexaFluor 488 and TRITC/Cy3/TagRFP/AlexaFluor 546. Universities and laboratories with applications in hematology, cytology and histology will look for this flexible microscope platform with a superb performance in optics and mechanics.
- Display maximum functionality and ergonomics in all its mechanical aspects
- Elegant and robust, the microscope stand follows all needs for an intense daily use
- Eyepiece tubes guarantee a fatigue-free work in terms of viewing angle and improved interpupillary distance
- The 'butterfly' movement may increase the viewing height by 60 mm for individual positioning
- CCIS© Infinity concept presents this apochromatic level of performance in a complete set of optional bright field objectives, compatible to all BA series models of upright microscopes
- Highest quality demands from pathology, histology and hematology can now be satisfied for a reliable diagnosis in routine and research work of hospitals and other biomedical labs
- Based on an encoded 6-fold nosepiece which memorizes the light intensity setting for each nosepiece position
- The system replicates this intensity setting once the respective position is swung in again; any new setting is taken as an overwriting of the former setup
Create a digital environment with Moticam: digitize your lab with our Moticam solutions. Look at your samples in your computer, your tablet or smartphone, a TV screen or a projector and view, capture, edit, measure, make reports or share your work with ease with anyone in just a few clicks. Our software solutions included in the packages show a user-friendly interface, making data sharing possible in a collaborative environment, encouraging teamwork and collective learning experiences.