HALO® PCS C18 HPLC Columns
Supplier: Advanced Materials Technology
New Product
Certificates
About this item
HALO® PCS C18 is a positively charged surface chemistry designed to deliver improved peak shapes for basic compounds. Ideal for use with low ionic strength mobile phases, HALO® PCS maintains peak symmetry at higher loading capacities and provides an alternate selectivity for other C18 bonded phases.
- Excellent peak shape and increased loading capacity for basic compounds
- UHPLC and LCMS compatible
- Alternate L1 selectivity
- Built on fused-core® technology for fast, efficient and reliable separations
Available in both a 90 Å for small molecule analysis and 160 Å pore size for peptide analysis.
Certifications: ISO 9001:2015 certified.
BodyTextAccessories: Use guard cartridges with guard holder.
Ordering information: Other dimensions columns and guard cartridges are available on request.
Delivery: Supplied with a test chromatogram and batch validation certificate.
Packaging: All packaging is recyclable.
Caution: These columns cannot be opened or repacked.
Specifications
- Catalog No:
- 77148-024
- 77148-032
- 77148-038
- 77148-036
- 77148-042
- 77148-040
- 77148-054
- 77148-026
- 77148-052
- 77148-030
- 77148-034
- 77148-028
- 77148-046
- 77148-044
- 77148-050
- 77148-048
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- pH range:
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- Temperature limit:
- 60...40 °C (Low to high)
- 60...40 °C (Low to high)
- 60...40 °C (Low to high)
- 60...40 °C (Low to high)
- 60...40 °C (Low to high)
- 60...40 °C (Low to high)
- 60...40 °C (Low to high)
- 60...40 °C (Low to high)
- 60...40 °C (Low to high)
- 60...40 °C (Low to high)
- 60...40 °C (Low to high)
- 60...40 °C (Low to high)
- 60...40 °C (Low to high)
- 60...40 °C (Low to high)
- 60...40 °C (Low to high)
- 60...40 °C (Low to high)
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
Specifications
Customers Who Bought This Also Bought