Fischertechnik TXT 4.0 Autonomous Driving Add-On Pack
Catalog #: 470332-594
Supplier: STUDICA INC.
Specifications
- Description:TXT 4.0 autonomous driving add-on pack
- Cat. no.:470332-594
Specifications
About this item
Allow students to discover a variety of exciting technologies essential to our everyday lives.
- Pair with the TXT 4.0 Base set
- Teaching Topics Include: autonomous driving, control technology, analog sensor technology, speed measurement, distance calculation, distance measurement
- Suitable for Middle/High School students
- Downloadable teaching and activity guides
Build and program your own 'car of the future' to amaze and delight the classroom! The Robotics Add On: Autonomous Driving set combines with the Robotics Base Set to allow students to discover a variety of exciting technologies essential to our everyday lives. From the automatic light to a lane departure warning system, cruise control and an automatic parking feature – the model guarantees excitement in the classroom. The building set includes a differential, as well as additional wheels, LEDs and a servo motor for the steering. The Robotics Add On: Autonomous Driving also includes seven experiments with associated solutions.
Ordering information: Required but available separately: TXT 4.0 Base set.
Customers Who Bought This Also Bought