mBot Ranger Robot Kit
Catalog #: 470324-126
Supplier: THE DOUGLAS STEWART CO.,INC
Specifications
- Description:mBot Ranger robot kit
- Cat. no.:470324-126
Specifications
About this item
This robot kit is a favorite in the classroom.
- Chassis: Aluminum alloy
- Processor: ATmega2560
- Coding language: Block-based or Arduino
- Communication mode: USB or Bluetooth
The mBot Ranger is an intermediate robotics kit designed for STEAM educators and robotics enthusiasts. Enjoy versatile hands-on activities in class, in workshops, at home, and more! It has three configurations that help children explore the mechanical engineering and design: robot tank, self-balancing robot, and racing car. Conquer all terrains like flat roads, grassland, gravel, and sandy beach.
When paired with the free programming apps and instructional resources, it provides a visual programming experience for all users. Supports Scratch and Arduino C programming, so kids can learn coding from beginner to expert.
Customers Who Bought This Also Bought