Macro Fluorescence Cuvette with PTFE Cover (Lightpaths: 1 - 100 mm), Type 1FL
Supplier: FIREFLYSCI, INC.
Certificates
About this item
Firefly's basic cuvette for fluorometric applications.
- Fire-fused to ensure that cuvettes are robust and durable
- Available in different materials: Optical glass, pyrex glass (10 mm only), UV quartz, IR quartz
- Lightpaths from 1 to 100 mm available
- Four windows and base optically polished
- Includes PTFE cover
This cell is the same build as the Type 1 with the only difference being that all four sides and the base are polished.
Customers Who Bought This Also Bought