Eisco Eudiometer Tubes
Supplier: Wards
Certificates
About this item
Eudiometer tubes with platinum elecrodes for measuring gas volume changes during chemical reactions.
- Made of mechanically strong and chemical resistant Borosilicate 3.3 glass
- Contains two platinum electrodes on the sealed end of tube
- Easy-to-read white graduations
Eudiometer tubes made of high quality Borosilicate glass. Useful for measuring volume of gas produced or consumed in a chemical reaction.
Customers Who Bought This Also Bought