To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Capacity:5 L (1.3 gal.)
- Description:Capped
- Cat. no.:77008-218
- Sterility:Sterile
Specifications
About this item
Stabilized hypochlorous acid blended with purified water.
- Filtered to 0.2 µm under ISO Class 5 airflow in a ISO Class 7 cleanroom ensure the product is free from contamination and particulates
- Contains no quaternary ammonium or surfactant very low residue, saving time on residue removal
- No hazard classification means good operator acceptability as no strong odor and only basic PPE is required
- Not classed as corrosive means it can be used safely in all areas of the cleanroom
Contec® ProKlean is a blend of hypochlorous acid in purified water. It is a ready- to-use cleaner safe enough for any surface in a cleanroom. Contec ProKlean is 0.2 µm filtered, and filled under Grade A unidirectional airflow in a Grade B/ISO Class 5 cleanroom. The sterile product is filled into pre-irradiated components. Provided double bagged, the product is designed for ease of entry into pharmaceutical cleanrooms. Ready-to-use so can be applied directly to the surface ensuring even coverage. Leave the surface wet for required contact time. Contec ProKlean is low residue but where residues are an issue they are easily removed with either water or alcohol.