Circular Specimen Chucks for Leica, TBS, and Tanner Cryostats, 55 mm, Available in Multiple Colors
Supplier: Mortech Manufacturing
New Product
Certificates
About this item
Available in multiple sizes and colors. Some variations also available in waffle pattern.
- Round, aluminum specimen chucks
- Suitable for Leica (except CM1950 model), TBS, and Tanner cryostats
- Some variations also available in waffle pattern
Available in red, orange, yellow, green, blue, purple, pink and white.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.