Vi-CELL BLU Startup Kit
Catalog #: CA76409-850
Supplier: Beckman Coulter
New Product
Specifications
- Description:Vi-CELL BLU startup kit, includes power supply (C09492), carousel assembly (C22467), power cord (873155), sample vials bag of 350 (C24843), 5×96 well plate (C99716), 10×96 well plate cover slip (C24842), allen wrench std hex 0.8 cm (0.31") (C17273), plug to cover locking screw hole (C17843)
Specifications
About this item
Vi-CELL BLU cell viability analyzer. The faster Vi-CELL BLU cell analyzer automate the trypan blue dye exclusion method for cell viability analysis.
- Fully automated sample preparation, analysis and post-run cleaning
- Small sample size
- Fast sample processing time
- Log-in samples on the fly
- Single sign-on with Active Directory
- Small laboratory footprint
- Flexible sample introduction-carousel or 96 well plate
- User-friendly reagent system
- Cell viability reported in percentage, concentration, and cell count
- Large sample capacity
- Data integrity tools for 21 CFR Part 11 compliance
The Vi-CELL BLU expedites processing by now having the option to use a 24-position sample carousel or a 96 well plate for sample delivery.
Caution: Safe for indoor use only.
Customers Who Bought This Also Bought