Portelite™ Fluorimetric Total Nucleic Acid Quantitation Kit
Catalog #: 76779-116
Supplier: AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Specifications
- Description:Kit is designed to rapidly measure the total amounts of nucleic acids, including double-stranded DNA (dsDNA), single-stranded DNA (ssDNA) and RNA in a sample
- Size:200 tests
Specifications
About this item
Portelite™ fluorimetric total nucleic acid quantitation kit is optimized for measuring the total amounts of nucleic acids with CytoCite™ or Qubit® fluorometers.
- Optimized for cytocite and qubit fluorometers
- Sensitive and indiscriminate binding
The kit has all the essential reagents, including Helixyte™ Green ssDNA reagent, dilution buffer, and pre-diluted DNA standards.
Packaging: Plastic vial.
Caution: For research use only.
Customers Who Bought This Also Bought