Portelite™ Fluorimetric ssDNA Quantitation Kit
Catalog #: 76779-046
Supplier: AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Specifications
- Description:Kit is designed to rapidly measure single-stranded DNA
- Size:200 tests
Specifications
About this item
The assay detects long oligonucleotides or ssDNA. Nucleotides and short oligo nucleotides of six bases or less do not interfere in the quantitation assay.
- Optimized for cytocite and qubit fluorometers
- Any volume from 1 to 20 µl is acceptable
- Assay range of 1 to 200 ng
The kit has all the essential reagents, including Helixyte™ Green ssDNA reagent, dilution buffer, and prediluted DNA standards. Helixyte™ Green ssDNA reagent is a sensitive fluorescent nucleic acid probe for quantifying oligonucleotides and single-stranded DNA (ssDNA) in solution.
Packaging: Plastic vial.
Caution: For research use only.
Customers Who Bought This Also Bought