Helixyte™ Green Fluorimetric ssDNA Quantitation Kit
Catalog #: 76779-058
Supplier: AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Specifications
- Description:Designed to measure single-stranded DNA in an easy and accurate format
- Size:200 tests
Specifications
About this item
Helixyte™ Green fluorimetric ssDNA quantitation kit is optimized for quantifying dsDNA with a fluorescence microplate reader.
- Optimized for microplate readers
- High sensitivity
- Low interference from common contaminants
The kit has all the essential reagents, including Helixyte™ Green ssDNA reagent, dilution buffer and prediluted DNA standards. Helixyte™ Green ssDNA reagent is a sensitive fluorescent nucleic acid probe for quantifying oligonucleotides and single-stranded DNA (ssDNA) in solution. It enables researchers to quantify as little as 100 pg/ml oligonucleotide or ssDNA. This sensitivity exceeds that achieved with absorbance methods by more than 10000-fold.
With this kit, as little as 1 ng/ml oligonucleotide or ssDNA might be detected.
Packaging: Plastic vial.
Caution: For research use only.
Customers Who Bought This Also Bought