Specifications
- Description:Red cabbage juice pH indicator
- Cat. no.:470334-898
- Includes:SDS and instructions
Specifications
About this item
A colorful way to test the pH of various solutions!
- Comes with 20 g of red cabbage powder and instructions
Learn about acids, bases, and pH indicators with our red cabbage juice kit. A pH indicator is a chemical that behaves differently depending on the pH level it is exposed to. Red cabbage juice is a natural pH indicator that changes color from red to purple to yellow-green as the pH of a solution increases. Test out different substances to determine if they are acidic or basic, and see if you can make all six colors found on the color chart!