About this item
Powerful digital disperser designed for the maximum level of reproducibility in a wide range of applications. Cutting-edge technology and engineering expertise guarantee precision and high shearing force for fast homogenisation, emulsification, suspensions, and high-speed mixing of biological tissues, pharmaceuticals, cosmetics and food products.
- Volume range from 0.2 ml to 2.5 L (H₂O), handles samples up to 5000 mPas viscosity
- The bright 3-digit LED display and the comfortable rotating knob enable precise electronic speed settings from 3000 to 25000 min⁻¹
- Various safety functions, such as a soft start, motor overload protection and overtemperature alarms ensure the highest safety in daily operations
- A resistant and robust housing protects the internal electronic parts and ensures the instrument works even in the most challenging applications
- Premium construction materials guarantee excellent mechanical properties and chemical resistance
- A wide range of dispersing tools ensures excellent performance for many applications for small samples and large volumes
- A fast and easy installation system enables to easily accommodate and change the tools
The OV 625 Digital incorporates cutting-edge technologies and high levels of engineering expertise, to guarantee high shearing force and excellent homogenizing, emulsifying, suspending and mixing performance for a wide range of applications. Thanks to a wide offer of dispersing tools, the OV 625 Digital enables processing volumes ranging from 0.2 millilitres to 2.5 litres (H₂O) and up to 5000 mPas viscous samples.
The resistant and rugged housing protects all internal electronic components and enables proper operations in the most challenging conditions. The ergonomic, compact and innovative design and the robust structure make the OV 625 Digital a solution designed to last, guaranteed by a 3-year warranty (online product registration required).
Choosing the proper dispersing tool is crucial to achieve the best performance and high reproducibility. A wide range of dispersing tools ensures excellent performance for many applications for small samples, sizes such as biological matrices, and large volumes (up to 4 litres as a high-speed mixer). The VELP dispersing tools are manufactured with premium construction materials to offer great compatibility, chemical resistance to various samples and reagents, and optimal mechanical properties. Moreover, a fast and easy installation system enables you to easily accommodate and change the tools, getting ready for the following applications in a breeze.
Specifications
- Catalog No:
- 77119-176
- 77119-174
- Volume H₂O:
- 0.2 - 2500 ml
- 0.2 - 2500 ml
- Max. viscosity:
- 5000 mPa
- 5000 mPa
- Speed range:
- 3000 - 25000 rpm
- 3000 - 25000 rpm
- Speed setting:
- 200 rpm
- 200 rpm
- W×D×H:
- 92×280×130 mm
- 92×280×130 mm
- Sound level:
- 75 dBa (without element)
- 75 dBa (without element)
- Material of construction:
- Aluminum, technopolymer
- Aluminum, technopolymer