SCIEX 7600 QTOF with Zeno Trap and EAD System
Catalog #: 76644-998
Supplier: AB SCIEX LLC CA
Specifications
- Description:QTOF with Zeno trap pulsing and EAD mass spectrometer
- Mass range:Isolating Quad 1:5 - 2250
TOF: 5 - 40,000
Dynamic range inter-scan: >5 orders of magnitude
Dynamic range intra-scan: 4 orders of magnitude - Scan:TOF MS, Q1, TOF MS/MS, MRMHR (with and without zeno), SWATH, Zeno SWATH, IDA
- Cat. no.:76644-998
Specifications
About this item
This high-resolution accurate mass system combines Zeno trap pulsing with EAD fragmentation (electron activated dissociation) to unlock sensitivity gains that reveal new, rare, or even previously undetected information. Detect up to 20× more ions in every experiment and access a spectrum of tunable fragmentation techniques to uncover new perspectives.
- Characterize large molecules including post-translational modifications
- Elucidate positional isomers on small molecules and lipids
- Identify and quantify proteins and peptides at unparalleled speed
- Gain significant sensitivity and discover less abundant ions
- EAD cell-tunable EAD between 0 to 25 eV
- Source: Turbo V ion source housing with Twinspray analytical probe for ESI or APCI; OptiFlow ion source housing with Optiflow micro and nano probes
A high-resolution mass spectrometry solution that combines powerful MS/MS sensitivity, fragmentation technology and a step-change in data independent acquisition.
Customers Who Bought This Also Bought