SCIEX X500R QTOF System
Catalog #: 76644-982
Supplier: AB SCIEX LLC CA
Specifications
- Description:X500R QTOF system, QTOF mass spectrometer
- Mass range:Isolating Quad 1:5 - 2250
TOF: 5 - 40,000
Dynamic range inter-scan: 4 orders of magnitude
Dynamic range intra-scan: 4 orders of magnitude - Scan:TOF MS, Q1, TOF MS/MS, MRMHR, SWATH, IDA
- Cat. no.:76644-982
Specifications
About this item
Engineered and versatile to handle the demands of any high throughput laboratory. Incorporating the robustness of turbo V ionization source along with MRMHR, it quantifies food and environmental testing samples to regulatory compliance requirements. For complete unknown screening, SWATH® acquisition detects any trace level compounds and delivers vital MS/MS data.
- Renowned SWATH Data Independent Acquisition (DIA) enables comprehensive detection, so you won’t miss any critical MS/MS information for your application needs
- SCIEX spectral libraries offer more compounds for more complete screening studies
- Gain efficiency, see improved usability and achieve greater integrity with easy to learn and use SCIEX OS software
- Source: TurboV ion source housing with Twinspray analytical probe for ESI or APCI
With simple workflows, robust hardware, sensitivity, and precision, the X500R QTOF is the first high-resolution LC-MS/MS system designed for both characterization and quantitation.
Customers Who Bought This Also Bought