SCIEX Triple Quad 7500 Systems
Supplier: AB SCIEX LLC CA
Certificates
About this item
The SCIEX 7500 system delivers a higher level of sensitivity than previous SCIEX systems. The system is integrated with innovative technology, which enables a low-level of quantification of analytes in crude and complex matrices.
- QTRAP activated and QTRAP ready options
- The D Jet ion guide concentrates samples and removes gas molecules and neutral ions
- Enhanced product ion scans enable improved confidence, while MRM3 workflows push quantification levels through matrix interferences
- Source: OptiFlow Pro ion source housing with OptiFlow Pro analytical probe and E lens for ESI
- Attain lower levels of quantification: The precise and robust engineering of the ion rail allows consistent and reproducible analysis time after time, by focusing the crucial ions you need for your workflow
Enable new levels of quantification using the SCIEX 7500 system, powered by SCIEX OS software.
The OptiFlow pro ion source introduces a new modularity feature and incorporates the reliability and efficiency of the legendary turbo V ion source.
Specifications
- Catalog No:
- 76644-996
- 76644-992
- Mass range:
- 5 - 2000, Triple Quad (dynamic range 6 orders of magnitude)
- 50 - 2000, QTRAP (dynamic range 6 orders of magnitude)
- Polarity switching:
- 5 ms
- 5 ms
- Scan:
- Full scan MS and selected ion for both Q1 and Q3, product ion scan, precursor ion scan, neutral loss or gain scan, multiple reaction monitoring (MRM)
- Enhanced MS scan, enhanced product ion scan, enhanced resolution scan, MS3 scan, MRM3 scan and TripleTrap scanning modes