OPTI-LYNX™ Holder Tube for 15 mm Guard Cartridge
Catalog #: 77100-360
Supplier: Optimize
Specifications
- Description:OPTI-LYNX™ holder tube for 15 mm guard cartridge
- Cat. no.:77100-360
- Supplier No.:11-02328-AA
Specifications
About this item
All OPTI-LYNX™ hardware can be purchased in kits or separately as needed for your application. OPTI-LYNX™ hardware is precision machined to maintain the highest quality and strictest tolerances.
- For use with OPTI-LYNX™ hardware
- Installs into holder cap to make connections with guard cartridges
- Modifies an OPTI-LYNX™ holder to accept guard cartridges
Installs into holder cap to make connections with OPTI-LYNX™ guard column cartridges (15 mm bed length).
Customers Who Bought This Also Bought