OPTI-LYNX™ Holder Tube for sealing element
Catalog #: 77100-358
Supplier: Optimize
Specifications
- Description:OPTI-LYNX™ holder tube for sealing element
- Cat. no.:77100-358
- Supplier No.:11-02327-AA
Specifications
About this item
All OPTI-LYNX™ hardware can be purchased in kits or separately as needed for your application. OPTI-LYNX™ hardware is precision machined to maintain the highest quality and strictest tolerances.
- For use with OPTI-LYNX™ hardware
- Installs into holder cap to make connections with sealing elements
- Modifies an OPTI-LYNX™ holder to accept sealing element inserts
Installs into holder cap to make connections with sealing elements.
Customers Who Bought This Also Bought