OPTI-LYNX™ Sealing Element
Catalog #: 77100-342
Supplier: Optimize
Specifications
- Description:OPTI-LYNX™ sealing element
- Cat. no.:77100-342
- Supplier No.:11-02307-AB
Specifications
About this item
All OPTI-LYNX™ hardware can be purchased in kits or separately as needed for your application. OPTI-LYNX™ hardware is precision machined to maintain the highest quality and strictest tolerances.
- For use with OPTI-LYNX™ hardware
- Provides a straight-through fluid connection when filters and packed beds are not required
Use an OPTI-LYNX™ direct-connect in-line kit & an extra bayonet connector to switch quickly and easily between multiple detectors or flow paths. Simply terminate the column outlet line with a holder cap/tube and use extra bayonet connectors for each detector inlet line.
Customers Who Bought This Also Bought