OPTI-LYNX™ Column Plug
Catalog #: 77100-340
Supplier: Optimize
Specifications
- Description:OPTI-LYNX™ column plug (fits bayonet OPTI-LYNX™ hardware)
- Cat. no.:77100-340
- Supplier No.:11-02293-AA
Specifications
About this item
Column plugs can be placed on OPTI-LYNX™ bayonet connectors to seal and protect fluid interfaces, packed beds, frits, and more.
- For use with OPTI-LYNX™ hardware
- Fits all OPTI-LYNX™ bayonet connectors
- Seals and protects fluid interfaces and filters/packed beds
Customers Who Bought This Also Bought