To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antigen name:SARS-CoV-2 RBD
- Clonality:Cocktail
- Isotype:IgG
- Neutralisation:Yes
- Conjugation:Unconjugated
- ELISA:Yes
- Size:20 μl
- Cross adsorption:No
- Form:Liquid
- Storage buffer:The SARS-CoV-2 Neutralizing Antibody Standard supplied in 0.2 μm filtered PBS, pH 7.4. The volume of this standard is 100 μl
- Storage temperature:This standard should be stored at −20 °C upon receipt until use. Avoid repeated freezing and thawing cycles
- Concentration:1,000,000 U/ml
- Immunogen:Recombinant SARS-CoV-2 RBD Protein
- Cat. no.:77007-644
- Packaging:Vial
Specifications
About this item
SARS-CoV-2 (Severe acute respiratory syndrome coronavirus 2), also known as SARS-CoV-2 (Severe acute respiratory syndrome coronavirus 2), also known as 2019-nCoV, is a positive-sense single-stranded RNA virus. It caused coronavirus disease in 2019 (COVID-19). SARS-CoV-2 contains glycosylated spike (S) protein, which is composed of S1 subunit and S2 subunit. The S1 contains a receptor-binding domain (RBD) that can bind to ACE2 receptor on target cells.Neutralizing antibodies against SARS-CoV-2 can block the interaction between SARS-CoV-2 RBD and ACE2. The blocking ability of SARS-CoV-2 neutralizing antibodies is affected by its quantity. This standard product is intended for the calibration of SARS-CoV-2 neutralizing antibodies in the materials or samples. It can be used in the assessment and development of assays for the detection and quantitation of SARS-CoV-2 neutralizing antibodies. The neutralizing antibody standard binds to multiple neutralizing epitopes in the receptor-binding domain. This product has been verified to calibrate the quantitative assay of SARS-CoV-2 neutralizing antibody with the Surrogate Virus Neutralization Test (sVNT) (GenScript, Cat.No L00847-A), plaque reduction neutralization test (PRNT) and pseudovirus neutralization test (pVNT) (GenScript, Cat.No SC2087). Users can design assays to detect and evaluate the quantitation of SARS-CoV-2 neutralizing antibodies by this product.
- High specificity
- High affinity
- High stability
Genscript offers site-specific Neutralizing antibodies, neutralizing antibody standards, and control antibodies.
Type:
Antigen: SARS-CoV-2 RBD
Clonality: Cocktail
Clone:
Conjugation:
Epitope:
Host:
Isotype: IgG
Reactivity: