Order Entry
Canada
ContactUsLinkComponent
Anti-AAV Rabbit Monoclonal Antibody [clone: 24F5]
Anti-AAV Rabbit Monoclonal Antibody [clone: 24F5]
  77007-634
 :  Genscript
Anti-AAV Rabbit Monoclonal Antibody [clone: 24F5]
  77007-634
 :  Genscript
 :  A02207-40
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

 

  • Antigen name:
    AAV
  • Clonality:
    Monoclonal
  • Clone:
    24F5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
  • Western blot:
    Yes
  • Size:
    40 μg
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Storage buffer:
    Lyophilized with PBS, pH 7.2, containing 0.02% sodium azide
  • Storage temperature:
    It is stable up to 1 year at −20 °C from date of receipt. The reconstituted antibody can be stored for 2 - 3 weeks at 2 - 8 °C. For long term storage, aliquot and store at −20 °C or below. Avoid repeated freezing and thawing cycles
  • Concentration:
    Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml
  • Immunogen:
    AAV8 Virus-Like Particles (VLPs)
  • Tested applications:
    Dot Blot
  • Cat. no.:
    77007-634
  • Packaging:
    Vial

 

 

Adeno-associated virus (AAV) is a single-stranded DNA virus that commonly infects humans without pathology. Therefore, AAV can be engineered to deliver DNA to target cells as a vector for gene therapy. There are several serotypes including AAV type 1, 2, 5, 6, 8, 9, DJ and rh.1. Among them, AAV2 is the best characterized and most commonly used. Cap gene of AAV gives rise to three capsid proteins, VP1, VP2 and VP3, with molecular weight of 87, 72 and 62 kDa respectively. These capsid proteins assemble into a near-spherical protein shell of 60 subunits for AAV.

  • High specificity
  • High affinity
  • High stability

GenScript's AAV antibody is a useful tool for evaluation of virus expression level, quantification of virus titer, identification of capsid proteins in AAV production, and analysis of AAV immunogenicity.

Type:
Antigen: AAV
Clonality: Monoclonal
Clone: 24F5
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: