Order Entry
ContactUsLinkComponent
Anti-Pertuzumab Mouse Monoclonal Antibody [clone: 11H4]
Anti-Pertuzumab Mouse Monoclonal Antibody [clone: 11H4]
Catalog #: 77007-500
Supplier:  Genscript
New Product
Anti-Pertuzumab Mouse Monoclonal Antibody [clone: 11H4]
Catalog #: 77007-500
Supplier:  Genscript
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antigen name:
    Pertuzumab
  • Clonality:
    Monoclonal
  • Clone:
    11H4
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Size:
    40 μg
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Storage buffer:
    This product is lyophilized with PBS, pH 7.2, containing 0.02% sodium azide
  • Storage temperature:
    The lyophilized product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2 - 8 °C or for up to 12 months at −20 °C or below. Avoid repeated freezing and thawing cycles
  • Concentration:
    Reconstitute the lyophilized antibody with deionized water (or equivalent) to a final concentration of 0.5 mg/ml
  • Immunogen:
    Pertuzumab
  • Cat. no.:
    77007-500
  • Packaging:
    Vial

Specifications

About this item

Pertuzumab (also called 2C4, trade name Perjeta) is a monoclonal antibody used in combination with trastuzumab and docetaxel for the treatment of metastatic HER2-positive breast cancer; it also used in the same combination as a neoadjuvant in early HER2-positive breast cancer. The epitope for pertuzumab is the domain of HER2 where it binds to HER3, and pertuzumab prevents the HER2/HER3 dimer from forming, which blocks signalling by the dimer to prevent HER2 from functioning. Anti-Pertuzumab antibody (11H4), mAb, Mouse is produced from a hybridoma resulting from the fusion of partner and B-lymphocytes obtained from a mouse immunized with Pertuzumab.

  • High specificity
  • High affinity
  • High stability

GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.

Type:
Antigen: Pertuzumab
Clonality: Monoclonal
Clone: 11H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:

Customers Who Bought This Also Bought