To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Human IgM mu Chain
- Clonality:Monoclonal
- Host:Mouse
- Isotype:IgG1
- Conjugation:Unconjugated
- ELISA:Yes
- Size:200 μl
- Cross adsorption:No
- Form:Liquid
- Storage buffer:50 mM Na-citrate, 150 mM NaCl, pH 7.0, containing 0.03% proclin 300
- Storage temperature:Store at −20 °C. This product is stable for 1 year upon receipt, when handled and stored as instructed. Avoid repeated freezing and thawing cycles
- Concentration:1 mg/ml
- Immunogen:Human IgM mu chain
- Cat. no.:77007-598
- Packaging:Vial
Specifications
About this item
Immunoglobulin M (IgM) is one of several isotypes of antibodies (also known as immunoglobulin) that are produced by vertebrates. IgM is the largest antibody, and it is the first antibody to appear in the response to initial exposure to an antigen. Formed from covalently linking 5 immunoglobulins together. Anti-human IgM mu chain antibody is ideal for investigators in Immunology, Microbiology, and Cell Biology.
- High specificity
- High affinity
- High stability
GenScript offers highly specific monoclonal and polyclonal primary antibodies. These primary antibodies are validated for multiple applications, including Sandwich ELISA and western blot.
Type: Primary
Antigen: Human IgM mu Chain
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: