To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antigen name:Sarilumab
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Conjugation:Unconjugated
- ELISA:Yes
- Size:40 μg
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:This product is lyophilized with PBS, pH 7.2, containing 0.02% sodium azide
- Storage temperature:The lyophilized product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2 - 8 °C or for up to 12 months at −20 °C or below. Avoid repeated freezing and thawing cycles
- Concentration:Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml
- Immunogen:Sarilumab
- Cat. no.:77007-534
- Packaging:Vial
Specifications
About this item
Sarilumab with the trade name of Kevzara, is a human monoclonal IgG1 antibody. It binds to membrane bound and soluble interleukin 6 (IL-6) receptor forms. It was FDA-approved in for the treatment of Rheumatoid Arthritis (RA) in combination with methotrexate. GenScript anti-sarilumab antibody, pAb, rabbit is produced from sera of a rabbit immunized with Sarilumab.
- High specificity
- High affinity
- High stability
GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.
Type:
Antigen: Sarilumab
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: