To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antigen name:Omalizumab
- Clonality:Monoclonal
- Clone:7G11
- Host:Mouse
- Isotype:IgG1 kappa
- Conjugation:Unconjugated
- ELISA:Yes
- Size:40 μg
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:This product is lyophilized with PBS, pH 7.2, containing 0.02% sodium azide
- Storage temperature:The lyophilized product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2 - 8 °C or for up to 12 months at −20 °C or below. Avoid repeated freezing and thawing cycles
- Concentration:Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml
- Immunogen:Omalizumab
- Packaging:Vial
Specifications
About this item
Omalizumab with trade name Xolair, is an FDA-approved drug for the treatment of moderate to severe persistent asthma in adults and adolescents (12 years of age and above). It is a recombinant humanized monoclonal antibody. It selectively binds to human immunoglobulin E (IgE). Omalizumab inhibits the binding of IgE to the high-affinity IgE receptor (FcεRI) on the surface of mast cells and basophils. Anti-Omalizumab Antibody (7G11), mAb, Mouse is produced from a hybridoma resulting from the fusion of partner and B-lymphocytes obtained from a mouse immunized with omalizumab.
- High specificity
- High affinity
- High stability
GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.
Type:
Antigen: Omalizumab
Clonality: Monoclonal
Clone: 7G11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: