Corning® Videodrop Device for Characterizing Nanoparticles, Corning
Catalog #: CA76809-050
Supplier: Corning
New Product
Specifications
- Description:Corning® Videodrop
- Cat. no.:CA76809-050
Specifications
About this item
The Corning® Videodrop utilizes the principles of interferometry in conjunction with microscopy to sense, detect, characterize, and track single nanoparticles in real time.
- Speed
- Under 1 minute loading, processing, and cleaning
- No calibration nor wash
- Repeatable and reproducible
- No inter-operator bias
- No parameter adjustment
The Corning® Videodrop utilizes the principles of interferometry in conjunction with microscopy to sense, detect, characterize, and track single nanoparticles in real time.
The Videodrop aligns with the Cell and Gene therapy segment of the bioproduction market due to the correlation of collecting physical titer of viral vectors such as Lentivirus, Retrovirus, Adenovirus, and Extracellular Vesicles in the range of 80 to 500 nm. Videodrop outputs are linear compared to p24 ELISA, RTqPCR, and NTA assays.
BodyTextAccessories: Videodrop Glass Slides are packaged 50 per case and are used in conjunction with the Corning Videodrop. The Videodrop sample holder is magnetic and connects to the stage to secure the glass slides for data analysis. The Videodrop audit trail option is required for GMP facilities. Videodrop protective cover is used when the unit is not in use. The LED protective cover is magnetic and placed over the LED light when the unit is not in use.
Ordering information: Accessories are sold separately.
Delivery: A Corning affiliate will set up the equipment once delivered.
Customers Who Bought This Also Bought