NOVAX® Class 00 Rubber Insulating Gloves with Straight Cuff, Black
Supplier: PIP
New Product
Certificates
About this item
Used exclusively for electrical purposes, such as power generation, electrical contractors, hybrid automotive repair/service, hybrid automobile manufacturing, industrial plant maintenance, utilities.
- Contoured shape reduces hand fatigue
- Silicone-free
- Available in 11" or 14" lengths
Natural rubber construction offers the required dielectric properties combined with flexibility, strength, and durability. Individually numbered for traceability and stamped with factory manufacturing date.
Certifications: Gloves meet ANSI/ASTM D120 standard and NFPA 70E for use around electrical hazards and arc flash protection.
Specifications
- Catalog No:
- 76944-430
- 76944-428
- 76944-414
- 76944-412
- 76944-442
- 76944-426
- 76944-424
- 76944-438
- 76944-440
- 76944-420
- 76944-422
- 76944-416
- 76944-418
- 76944-436
- 76944-432
- 76944-434
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Cuff style:
- Straight
- Straight
- Straight
- Straight
- Straight
- Straight
- Straight
- Straight
- Straight
- Straight
- Straight
- Straight
- Straight
- Straight
- Straight
- Straight
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- ASTM No.:
- ASTM D120-14a
- ASTM D120-14a
- ASTM D120-14a
- ASTM D120-14a
- ASTM D120-14a
- ASTM D120-14a
- ASTM D120-14a
- ASTM D120-14a
- ASTM D120-14a
- ASTM D120-14a
- ASTM D120-14a
- ASTM D120-14a
- ASTM D120-14a
- ASTM D120-14a
- ASTM D120-14a
- ASTM D120-14a
Specifications
Customers Who Bought This Also Bought