Specifications
- Description:PTS Connect® printer
- Storage Temperature:20...30 °C (68...86 °F)
Specifications
About this item
The PTS Connect® Printer easily connects to the CardioChek® PA or CardioChek® Plus analyzers via a provided cable, and one roll of labels is provided with the purchase of the printer.
The PTS Connect® printer is designed to output biometric information on paper and self-adhesive labels. The printer output presents the biometric information in an easy-to-read format for medical professionals and patients.