Specifications
- Cat. no.:76994-114
- Storage Temperature:18...28 °C (64...82 °F) for a cumulative total of 4 months
Specifications
About this item
The A1CNow®+ system provides healthcare professionals with a fast and easy way of obtaining accurate A1C results with just a fingerstick.
- More efficient than the lab
- Results in 5 minutes
- Small (5 µl) blood sample
- Portable – use in multiple exam rooms
- Easy to use – minimal training required
- No maintenance
- Available with IFCC (mmol/mol), Measuring ranges; 4 to 13% A1C
A1CNow+ test system provides quantitative measurement of the percent of glycated hemoglobin (% A1C) levels in capillary fingerstick or venous whole blood samples. The test is for professional use to monitor glycemic control in people with diabetes. The test takes only 5 minutes and tells you how well the patient has been controlling their blood sugar over the past 2 to 3 months.
A1CNOW®+ LAB QUALITY RESULTS AT THE POINT OF CARE. The A1CNow®+ system provides healthcare professionals with a fast and easy way of obtaining accurate A1C results with just a fingerstick. This innovative technology enables clinicians to communicate face-to-face with patients about their diabetes control in minutes, not days. The analyzer, test cartridges, and shaker kit pouches may be used until the expiration date printed on the box when stored refrigerated at 2 to 8 °C (36 to 46 °F).