VWR® Deep Well Plates, 96-Well and 384-Well
Supplier: SCIENTIFIC SPECIALTIES, INC.
New Product
Certificates
About this item
VWR® DeepWell plates are ideal for sample collection, preparation, and long-term storage.
- Centrifugation to 4000×g
- Manufactured from virgin polypropylene to ensure purity and sample integrity, chemical resistance, and compatibility with long-term storage needs
- Alphanumeric indexing molded in on A-H and 1-12 axes
- Optimum sample mixing and recovery with highly-polished wells and radiused corners
- Raised rims on each well facilitate secure sealing with films and sealing mats
- Bar coded plates available—please inquire
VWR® 96- and 384-well deep well plates are HTS-friendly and compatible with most liquid-handling robots. Plates are manufactured from virgin polypropylene to ensure purity and sample integrity, chemical resistance, and compatibility with long-term storage needs.
Certifications: Certified free of detectable DNase, RNase, DNA, and PCR inhibitors. Sterile plates are also endotoxin-free.
Customers Who Bought This Also Bought