Specifications
- Description:Version 103 110V SteraMist Surface Unit
Specifications
About this item
The SteraMist Surface Unit is a portable, handheld point-and-spray disinfection and decontamination system.
- Mobile for rapid deployment throughout a variety of spaces
- Economical, non-corrosive, and easy to apply
- Easily incorporated into daily cleaning protocols
- Portable and rugged design for easy storage
- 30’ removable applicator cord helps you reach a wide radius
- Routine disinfection for large areas and surfaces
The SteraMist Surface Unit is the perfect solution for facility surface disinfection, bringing you handheld control with more movement and mobility around the treated area.