SteraMist Total Disinfection Cart
Catalog #: 76919-434
Supplier: TOMI ENVIRONMENTAL SOLUTIONS, INC
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Stainless Steel Cart with Version 103 110V SteraMist Surface Unit with and accessories
Specifications
About this item
The SteraMist Disinfection Cart utilizes the SteraMist Surface Unit in a convenient hospital-grade cart.
Delivery: Includes SteraMist® Surface Unit (SUR-103-110), durable shelved stainless steel transport cart, 2 gal. of BIT solution, Dräger® X-am 5100 Portable H2O2 Monitor, X-POWER 3580 air scrubber with dual carbon filters, MaxAir® system helmet with positive pressure air flow system, caution sign.
Customers Who Bought This Also Bought