SteraMist BIT Solution
Supplier: TOMI ENVIRONMENTAL SOLUTIONS, INC
New Product
Certificates
About this item
- All SteraMist products exclusively use low-percentage 7.8% Hydrogen Peroxide as a sole active ingredient
- Each SteraMist product utilizes the same BIT™ Solution blend for a wide array of applications
- BIT Solution kills within seconds of contact
- Non-corrosive to enure superior surface compatibility
- No toxic byproducts
- No mixing or dilution
- No wiping or rinsing
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.