SteraMist Environment System
Catalog #: 76919-446
Supplier: TOMI ENVIRONMENTAL SOLUTIONS, INC
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:SteraMist Environment System - 110/120 V includes three remote applicators, three stands and mounts, one tool kit, one DI water cartridge
Specifications
About this item
The SteraMist Environment System, the most intelligent mobile unit, delivers disinfection and decontamination with unparalleled scalability.
- 3 removable 30’ cords and applicator stands
- Intuitive HMI and custom cycle creation to meet all GMP requirements
- Features remote operation and hand-held modes
- Contains applicators able to treat areas of any size
Ideal for outbreaks and severe decontamination and deodorizing needs.
Delivery: Includes 3× remote applicators (single point connectors with 30’ cords), 3× remote applicator stands and mounts, tool kit, 1 gal. DI water cartridge.
Customers Who Bought This Also Bought