Somatic Cell Count Microscope Slide
Catalog #: 76357-804
Supplier: Bellco Glass
Specifications
- Description:Somatic cell count slide
- L×W:75×25 mm
- Cat. no.:76357-804
- Packaging:Case of 144
Specifications
About this item
The somatic Cell Count Slide is useful for direct microscopic counts of somatic cells in cows milk.
Spread 0.01 ml sample onto each circle than dry and stain. Count cells under oil immersion microscopy.
Customers Who Bought This Also Bought